Learn More
Aviva Systems Biology Corp P2RX2 Antibody - middle region (ARP35259_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35259_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against P2RX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human P2RX2. Alternate names: P2X2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55684