Learn More
Aviva Systems Biology Corp P2rx2 Antibody - middle region (ARP35437_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2rx2. It was validated on Western Blot. . Alternate names: MGC129148, MGC129149, P2X2a, P2x2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-463