Learn More
Aviva Systems Biology Corp P2RX7 Antibody - middle region (ARP35112_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35112_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2RX7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human P2RX7. Alternate names: MGC20089, P2X7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55646