Learn More
Eurofins Discoverx ChemiScreen P2Y1 Purinergic Receptor Stable Cell Line, 2 vials. (HTS049C)
Catalog No. 502005752
Encompass
The ChemiScreen P2Y1 Purinergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ADP, ATP) based activation of P2RY1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses P2RY1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of P2RY1, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5752 Supplier Eurofins Discoverx Supplier No. HTS049C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen P2Y1 Purinergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ADP, ATP) based activation of P2RY1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses P2RY1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of P2RY1, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.