Learn More
Eurofins Discoverx PathHunter P2RY12 Total Internalization U2OS Cell Line, 2 vials. (93-0952C3)
Catalog No. 502001652
Encompass
The PathHunter P2RY12 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of P2RY12 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged P2RY12 and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the P2RY12 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1652 Supplier Eurofins Discoverx Supplier No. 930952C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter P2RY12 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of P2RY12 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged P2RY12 and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the P2RY12 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.