Learn More
Aviva Systems Biology Corp P2RY13 Antibody - C-terminal region (ARP64924_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64924_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2RY13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human P2RY13. Alternate names: FKSG77, GPCR1, GPR86, GPR94, P2Y13, SP174ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-393