Learn More
Aviva Systems Biology Corp P2RY14 Antibody - C-terminal region (ARP65187_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65187_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2RY14. It was validated on Western Blot. . Alternate names: GPR105, KIAA0001, P2Y14ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-452