Learn More
Aviva Systems Biology Corp P2RY2 Antibody - C-terminal region (ARP63242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P2RY2. It was validated on Western Blot. . Alternate names: HP2U, MGC20088, MGC40010, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2RApplicationWB. Species reactivity: Human
Catalog No. 50-287-919