Learn More
Cusabio Technology LLC Human P2X purinoceptor 7(P2RX7) ELISA kit
Catalog No. NC2952981
Encompass_Preferred
This Human P2RX7 ELISA Kit was designed for the quantitative measurement of Human P2RX7 protein in serum, plasma, tissue homogenates, cell lysates. It is a Sandwich ELISA kit, its detection range is 25 pg/mL-1600 pg/mL and the sensitivity is 6.25 pg/mL. One kit contains 96 wells.
Catalog No. NC2952981 Supplier Cusabio Technology LLC Supplier No. CSBEL017325HU
This Human P2RX7 ELISA Kit was designed for the quantitative measurement of Human P2RX7 protein in serum, plasma, tissue homogenates, cell lysates. It is a Sandwich ELISA kit, its detection range is 25 pg/mL-1600 pg/mL and the sensitivity is 6.25 pg/mL. One kit contains 96 wells.