Learn More
Enzo Life Sciences Purinergic receptor P2X4 (mouse) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX215033R100
Encompass_Preferred
Host: Rabbit. Immunogen: Vector containing the cDNA of mouse purinergic receptor P2X4. Species reactivity: Mouse. Applications: Flow Cytometry, ICC, IP. Recommended Dilutions: Flow Cytometry (1:100-1:400) Immunocytochemistry (1:100-1:400) Immunoprecipitation (1:100-1:400) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Antiserum diluted with PBS, containing 0.1% BSA and 0.02% sodium azide.
Catalog No. 50-200-8203