Learn More
Santa Cruz Biotechnology P2X7 CRISPR/Cas9 KO Plasmid, human-specific, 20 μg, transfection-ready, GFP selection marker
Catalog No. NC3347157
Encompass
The P2X7 CRISPR/Cas9 KO Plasmid is a specialized tool designed for efficient and permanent knockout of the human P2X7 receptor gene. Utilizing the CRISPR/Cas9 mechanism, this plasmid offers a precise method for gene disruption in human cell lines to study physiological roles and pathological implications. Supplied as transfection-ready purified plasmid DNA, it streamlines experimental workflows for genetic manipulation studies.
  • Target-specific gene knockout ensures specific genetic disruption.
  • High knockout efficiency with a pool of three plasmids containing Cas9 nuclease and P2X7-specific gRNA.
  • Validated gRNA sequences from the GeCKO (v2) library enable reliable gene targeting.
  • Site-specific double-strand breaks (DSBs) lead to permanent gene disruption.
  • Transfection-ready format simplifies experimental setup.
  • GFP marker allows monitoring of transfection efficiency.
Catalog No. NC3347157 Supplier Santa Cruz Biotechnology Supplier No. SC400780
The P2X7 CRISPR/Cas9 KO Plasmid is a specialized tool designed for efficient and permanent knockout of the human P2X7 receptor gene. Utilizing the CRISPR/Cas9 mechanism, this plasmid offers a precise method for gene disruption in human cell lines to study physiological roles and pathological implications. Supplied as transfection-ready purified plasmid DNA, it streamlines experimental workflows for genetic manipulation studies.
  • Target-specific gene knockout ensures specific genetic disruption.
  • High knockout efficiency with a pool of three plasmids containing Cas9 nuclease and P2X7-specific gRNA.
  • Validated gRNA sequences from the GeCKO (v2) library enable reliable gene targeting.
  • Site-specific double-strand breaks (DSBs) lead to permanent gene disruption.
  • Transfection-ready format simplifies experimental setup.
  • GFP marker allows monitoring of transfection efficiency.