Learn More
Santa Cruz Biotechnology P2X7 HDR Plasmid (h), 20 μg, Human, Puromycin resistance gene, RFP gene, Pool of 2-3 plasmids, Homology arms: 2 x 800 bp, 0.1 μg/μL
Catalog No. NC3351470
Encompass
P2X7 HDR Plasmid (h), 20 μg, Human, Puromycin resistance gene, RFP gene, Pool of 2-3 plasmids, Homology arms: 2 x 800 bp, 0.1 μg/μL
Catalog No. NC3351470 Supplier Santa Cruz Biotechnology Supplier No. SC400780HDR
P2X7 HDR Plasmid (h), 20 μg, Human, Puromycin resistance gene, RFP gene, Pool of 2-3 plasmids, Homology arms: 2 x 800 bp, 0.1 μg/μL