Learn More
Enzo Life Sciences Purinergic receptor P2X7 (mouse) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX215035R100
Encompass_Preferred
Host: Rabbit. Immunogen: Vector containing the cDNA of mouse purinergic receptor P2X7. Species reactivity: Mouse. Applications: Flow Cytometry, ICC, IP, FUNC. Formulation: Liquid. Antiserum diluted with PBS containing 0.1% BSA and 0.02% sodium azide.
Catalog No. 50-200-8205