Learn More
Enzo Life Sciences Purinergic receptor P2X7 (mouse) monoclonal antibody (HANO43) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX802027L001
Encompass_Preferred
Clone: HANO43. Isotype: IgG2b. Immunogen: Vector containing the cDNA of mouse purinergic receptor P2X7. Species reactivity: Mouse. Applications: Flow Cytometry, ICC, IP. Formulation: Liquid. Hybridoma supernatant containing 0.02% sodium azide.
Catalog No. 50-200-9722