Learn More
Ted Pella Inc P3 OIL FOR # 7010 ROTARY PUMP
SDP
Catalog No. NC0269652
Encompass
Clear, monograde oil that has the correct viscosity for Pfeiffer Rotary vacuum pumps, excellent sealing and lubrication properties, low vapor pressure and reduced back streaming. Used also for Prod. No. 7010 and 7010-220 Rotary Pumps. Indefinite shelf life when stored in unopened container at ambient temperatures out of direct sunlight. 1L
Catalog No. NC0269652 Supplier Ted Pella Inc Supplier No. 89138
Clear, monograde oil that has the correct viscosity for Pfeiffer Rotary vacuum pumps, excellent sealing and lubrication properties, low vapor pressure and reduced back streaming. Used also for Prod. No. 7010 and 7010-220 Rotary Pumps. Indefinite shelf life when stored in unopened container at ambient temperatures out of direct sunlight. 1L