Learn More
Lonza Walkersville P3 Primary Cell 96-well Nucleofector Kit (960 RCT)
Catalog No. NC1771278
Encompass
The P3 Primary Cell 96-well Nucleofector Kit is one of our five kits suited for medium-throughput transfection of primary cells when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies for primary cellsCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D-Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:22.5 mL P3 Primary Cell Nucleofector Solution5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 96-well Nucleocuvette Plate (20 μl)
Catalog No. NC1771278 Supplier Lonza Walkersville Supplier No. V4SP3960
Add to Cart
Add to Cart
Request a Quote
The P3 Primary Cell 96-well Nucleofector Kit is one of our five kits suited for medium-throughput transfection of primary cells when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies for primary cellsCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D-Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:22.5 mL P3 Primary Cell Nucleofector Solution5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 96-well Nucleocuvette Plate (20 μl)