Learn More
Azzota Corp P3 Smart Pump EZ 1
Catalog No. 5000023510
Encompass_Preferred
P3- PeriScientific Smart Pump Systems 0 007-3100 mL/min single head pump EZ 1
Catalog No. 50-000-23510 Supplier Azzota Corp Supplier No. PERIP3EZA1
P3- PeriScientific Smart Pump Systems 0 007-3100 mL/min single head pump EZ 1