Learn More
AUTOMATION TRAINER P30, Clear, Unfiltered, Sterile, Racked (50 384-Tip Racks/Case)
SDP
Catalog No. NC2044899
Encompass_Preferred
Automation Trainer's Beckman-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Beckman's 384-Format P30 & P30XL tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required. Maximum working volumes are 30 L for the Unfiltered P30 Tip and 50 L for the Unfiltered P30XL Tip.
Catalog No. NC2044899 Supplier AUTOMATION TRAINER Supplier No. BP30CLUSR384
Automation Trainer's Beckman-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Beckman's 384-Format P30 & P30XL tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required. Maximum working volumes are 30 L for the Unfiltered P30 Tip and 50 L for the Unfiltered P30XL Tip.