Learn More
Enzo Life Sciences P32/98 (50mg). CAS: 136259-20-6
SDP
Supplier:  Enzo Life Sciences BMLPI1420050
Encompass_Preferred
P32/98 (Ile-thiazolidide) is a specific, competitive transition-state substrate analog inhibitor of dipeptidyl peptidase IV (DPIV; DPPIV; CD26), with a Ki of 130 nM. Useful in diabetes research. Has been used in vivo and in tissue culture. Alternative name: 3-N- ((2S 3S)-2-Amino-3-methylpentanoyl -1 3-thiazolidine hemi-fumarate. Purity: ≥98% (TLC, free base). Solubility: Soluble in water (20mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1481