Learn More
TARGETMOL CHEMICALS INC P32/98 HEMIFUMARATE 5MG
SDP
Catalog No. 502577199
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. P32/98 hemifumarate is a dipeptidyl peptidase IV (DPP4) inhibitor with hypoglycemic properties that improves insulin sensitivity and β-cell responsiveness in the Zucker diabetic rat model and may be useful in studies of type 2 diabetes and colitis. purity: 99%
Catalog No. 50-257-7199 Supplier TARGETMOL CHEMICALS INC Supplier No. T219795MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. P32/98 hemifumarate is a dipeptidyl peptidase IV (DPP4) inhibitor with hypoglycemic properties that improves insulin sensitivity and β-cell responsiveness in the Zucker diabetic rat model and may be useful in studies of type 2 diabetes and colitis. purity: 99%