Learn More
Medchemexpress LLC P32 inhibitor M36 | 802555-85-7 | MFCD31666371 | 98.6% | 448.52 g·mol-1 | C23H28N8O2 | 25MG
SDP
Catalog No. 5000235843
Encompass_Preferred
p32 inhibitor M36 is a small-molecule research reagent that binds directly to the mitochondrial p32 (C1QBP) protein and blocks its association with LyP-1, enabling biochemical and cell-based studies investigating p32-related pathways such as proliferation and viability.
  • Binds directly to mitochondrial p32 protein.
  • Inhibits p32 association with LyP-1.
  • Suitable for cell-based and biochemical assays.
  • High purity for research applications.
  • Supplied as a solid, convenient for storage and handling.
  • Molecular formula C23H28N8O2; molecular weight 448.52 g·mol-1.
Catalog No. 50-002-35843 Supplier Medchemexpress LLC Supplier No. HY12471825MG
p32 inhibitor M36 is a small-molecule research reagent that binds directly to the mitochondrial p32 (C1QBP) protein and blocks its association with LyP-1, enabling biochemical and cell-based studies investigating p32-related pathways such as proliferation and viability.
  • Binds directly to mitochondrial p32 protein.
  • Inhibits p32 association with LyP-1.
  • Suitable for cell-based and biochemical assays.
  • High purity for research applications.
  • Supplied as a solid, convenient for storage and handling.
  • Molecular formula C23H28N8O2; molecular weight 448.52 g·mol-1.