Learn More
Abcam P32/98, dipeptidyl peptidase IV (DPP4) inhibitor, 10MG
Catalog No. 502706468
Encompass_Preferred
MW 318.39 Da, Purity >98%. Potent, selective dipeptidyl peptidase IV (DPP4) inhibitor (IC₅₀ = 131 nM). Orally active. Enhances glucose tolerance through increased insulin secretion. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6468 Supplier Abcam Supplier No. AB14172410MG
MW 318.39 Da, Purity >98%. Potent, selective dipeptidyl peptidase IV (DPP4) inhibitor (IC₅₀ = 131 nM). Orally active. Enhances glucose tolerance through increased insulin secretion. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement