Learn More
Cayman Chemical P3298hemIfumarate 10mg
SDP
Catalog No. 502966898
Encompass_Preferred
A competitive transition-state substrate analog inhibitor of DPP IV (Ki = 126 nM); used to improve glucose tolerance, insulin sensitivity, and β-cell glucose responsiveness in diabetic rat models at 20 mg/kg/day
Catalog No. 50-296-6898 Supplier Cayman Chemical Supplier No. 1688710 MG
A competitive transition-state substrate analog inhibitor of DPP IV (Ki = 126 nM); used to improve glucose tolerance, insulin sensitivity, and β-cell glucose responsiveness in diabetic rat models at 20 mg/kg/day