Learn More
Sino Biological Recombinant Human p38 Protein (Full Length, GST Tag), Active
Catalog No. 502568607
Encompass_Preferred
Recombinant full length human p38 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-256-8607 Supplier Sino Biological Supplier No. M3910BG10
Recombinant full length human p38 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.