Learn More
TARGETMOL CHEMICALS INC p38-alpha MAPK-IN-1 5MG
SDP
Catalog No. 502878418
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.p38-alpha MAPK-IN-1 is a MAPK14 (p38-alpha) inhibitor with an IC50 of 2300 nM in the EFC displacement assay and 5500 nM in the HTRF assay. Purity 99.52%
Catalog No. 50-287-8418 Supplier TARGETMOL CHEMICALS INC Supplier No. T123475MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.p38-alpha MAPK-IN-1 is a MAPK14 (p38-alpha) inhibitor with an IC50 of 2300 nM in the EFC displacement assay and 5500 nM in the HTRF assay. Purity 99.52%