Learn More
Sino Biological Recombinant Human P38 Gamma/MAPK12 Protein (Full Length, GST Tag), Active
Catalog No. 502574273
Encompass_Preferred
Recombinant full length human p38 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4273 Supplier Sino Biological Supplier No. M3710BG50
Recombinant full length human p38 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.