Learn More
Sino Biological Recombinant Human JNK2 Protein (Full Length, GST Tag), Active
Catalog No. 502574294
Encompass_Preferred
Recombinant full length human JNK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4294 Supplier Sino Biological Supplier No. M3410BG500
Recombinant full length human JNK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.