Learn More
Enzo Life Sciences p38 (pThr180/Tyr182) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900101
Encompass_Preferred
The [pThr180/Tyr182]p38 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Use less precious sample per test compared to Western blot with higher throughput sample analysis; measure up to 40 samples in duplicate per kit. Alternative name: SAPK2, SAP kinase 2. Sensitivity: 52.1 pg/ml (range 156 - 5,000 pg/ml). Use/Stability: Store all components at 4°C, except standard at -20°C. UniProt ID: Q16539 (human p38α), Q15759 (human p38β), P53778 (p38γ), O15264 (p38δ). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human, Mouse . Content: Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.
Catalog No. 50-200-6843