Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p38 gamma (MAPK12) protein, human | >95.0% | 50 UG
SDP

Catalog No. 5000290106
Encompass_Preferred

Recombinant, tag-free human p38 gamma (MAPK12) protein supplied as a filtered buffered solution for biochemical and cell-based research. The protein is full-length and prepared to support kinase-related assays and binding studies while minimizing contaminants and endotoxin.

  • Full-length human sequence (M1-L367).
  • Expressed in Sf9 insect cells to support eukaryotic folding and modifications.
  • Tag free to avoid interference in interaction studies.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Supplied in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0 (0.22 μm filtered solution).
  • Endotoxin <1 EU/μg, determined by LAL.
  • Shipped on dry ice and recommended storage at -80°C for long-term stability.

Catalog No. 50-002-90106 Supplier Medchemexpress LLC Supplier No. HYP7653250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, tag-free human p38 gamma (MAPK12) protein supplied as a filtered buffered solution for biochemical and cell-based research. The protein is full-length and prepared to support kinase-related assays and binding studies while minimizing contaminants and endotoxin.

  • Full-length human sequence (M1-L367).
  • Expressed in Sf9 insect cells to support eukaryotic folding and modifications.
  • Tag free to avoid interference in interaction studies.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Supplied in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0 (0.22 μm filtered solution).
  • Endotoxin <1 EU/μg, determined by LAL.
  • Shipped on dry ice and recommended storage at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.