Learn More
Medchemexpress LLC HY-131335 5mg Medchemexpress, p38α inhibitor 2 CAS:1095003-80-7 Purity:>98%
SDP
Catalog No. 502081273
Encompass_Preferred
Medchemexpress, HY-131335 5mg p38α inhibitor 2 CAS:1095003-80-7 p38α inhibitor 2 is a highly potent and selective p38α MAPK inhibitor, with a pIC50 of 9.6. p38α inhibitor 2 inhibits the hERG ion channel (IC50=27 μM) and shows a promising selectivity profile when tested in a panel of 51 other protein kinases (<30% inhibition at 10 μM concentration) and a panel of 141 other biological targets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1273 Supplier Medchemexpress LLC Supplier No. HY1313355MG
Medchemexpress, HY-131335 5mg p38α inhibitor 2 CAS:1095003-80-7 p38α inhibitor 2 is a highly potent and selective p38α MAPK inhibitor, with a pIC50 of 9.6. p38α inhibitor 2 inhibits the hERG ion channel (IC50=27 μM) and shows a promising selectivity profile when tested in a panel of 51 other protein kinases (<30% inhibition at 10 μM concentration) and a panel of 141 other biological targets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.