Learn More
Bioss p38 MAP Kinase - C-terminal M
SDP
Catalog No. 5000065669
Encompass_Preferred
p38 MAP Kinase (C-terminal) M138 Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M138Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65669 Supplier Bioss Supplier No. BSM70583M
p38 MAP Kinase (C-terminal) M138 Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M138Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol