Learn More
Abcam p38 MAPK alpha (pT180/Y182) ELISA Kit, 1 x 96 Tests
Catalog No. 502652218
Encompass_Preferred
p38 MAPK alpha (pT180/Y182) ELISA Kit is a single-wash 90-min Simplestep used to quantify p38 MAPK alpha (pT180/Y182) with a sensitivity of 200 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-2218 Supplier Abcam Supplier No. AB2210111X96TESTS
Add to Cart
Add to Cart
p38 MAPK alpha (pT180/Y182) ELISA Kit is a single-wash 90-min Simplestep used to quantify p38 MAPK alpha (pT180/Y182) with a sensitivity of 200 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement