Learn More
Medchemexpress LLC HY-18874 1mg Medchemexpress, p38-α MAPK-IN-1 CAS:443913-15-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY188741MG
Encompass_Preferred
Medchemexpress, HY-18874 1mg p38-α MAPK-IN-1 CAS:443913-15-3 p38-α MAPK-IN-1 is an inhibitor of MAPK14 (p38-α), with IC50 of 2300 nM in EFC displacement assay, and 5500 nM in HTRF assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2090