Learn More
Medchemexpress LLC P38-α MAPK-IN-1 | 443913-15-3 | 99.7% | 477.60 | 5 MG
SDP
Catalog No. 5000385236
Encompass_Preferred
p38-α MAPK-IN-1 is an inhibitor of MAPK14 (p38-α) that demonstrates inhibitory activity in both EFC displacement and HTRF assays. It also effectively inhibits LPS-induced tumor necrosis factor-alpha (TNF-alpha) production in THP-1 cells. This product is intended for research use only.
  • Inhibits MAPK14 (p38-α)
  • Effective in EFC displacement and HTRF assays
  • Inhibits LPS-induced tumor necrosis factor-alpha (TNF-alpha) production
Catalog No. 50-003-85236 Supplier Medchemexpress LLC Supplier No. HY188745MG
p38-α MAPK-IN-1 is an inhibitor of MAPK14 (p38-α) that demonstrates inhibitory activity in both EFC displacement and HTRF assays. It also effectively inhibits LPS-induced tumor necrosis factor-alpha (TNF-alpha) production in THP-1 cells. This product is intended for research use only.
  • Inhibits MAPK14 (p38-α)
  • Effective in EFC displacement and HTRF assays
  • Inhibits LPS-induced tumor necrosis factor-alpha (TNF-alpha) production