Learn More
TARGETMOL CHEMICALS INC P38 MAPK INHIBITOR 5MG
SDP
Catalog No. 502580200
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. p38 MAPK inhibitor is a potent inhibitor of p38 MAP kinase (IC50 = 35 nM). It inhibits senescence induced by the oncogene RAS. purity: 99%
Catalog No. 50-258-0200 Supplier TARGETMOL CHEMICALS INC Supplier No. T360105MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. p38 MAPK inhibitor is a potent inhibitor of p38 MAP kinase (IC50 = 35 nM). It inhibits senescence induced by the oncogene RAS. purity: 99%