Learn More
Cayman Chemical p38 MAPK InhIbItr 1mg
SDP
Catalog No. 502980085
Encompass_Preferred
A potent inhibitor of p38 MAP kinase (IC50 = 35 nM)
Catalog No. 50-298-0085 Supplier Cayman Chemical Supplier No. 176501 MG
A potent inhibitor of p38 MAP kinase (IC50 = 35 nM)