Learn More
Cayman Chemical p38 MAPK InhIbItr 500ug
SDP
Catalog No. 502966718
Encompass_Preferred
A potent inhibitor of p38 MAP kinase (IC50 = 35 nM)
Catalog No. 50-296-6718 Supplier Cayman Chemical Supplier No. 17650500 UG
A potent inhibitor of p38 MAP kinase (IC50 = 35 nM)