Learn More
Cayman Chemical p38 MAPK InhIbItr IV 5mg
SDP
Catalog No. 502987845
Encompass_Preferred
An ATP-competitive inhibitor of p38 MAPKs; inhibits p38α, p38β, p38��, and p38δ MAPKs (IC50s = 0.13, 0.55, 5.47, and 8.63 μM, respectively); inhibits LPS-induced TNF-α and IL-1β cytokine production in human peripheral blood mononuclear cells (IC50s = 22 and 44 nM, respectively)
Catalog No. 50-298-7845 Supplier Cayman Chemical Supplier No. 222195 MG
An ATP-competitive inhibitor of p38 MAPKs; inhibits p38α, p38β, p38��, and p38δ MAPKs (IC50s = 0.13, 0.55, 5.47, and 8.63 μM, respectively); inhibits LPS-induced TNF-α and IL-1β cytokine production in human peripheral blood mononuclear cells (IC50s = 22 and 44 nM, respectively)