Learn More
Cayman Chemical p38 MAPK PhosphoThr180 Tyr18
SDP
Catalog No. 502886724
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Thr180 and phospho-Tyr182 of rat p38 MAPK • Host: rabbit • Cross Reactivity: (+) human p38 MAPK • Application(s): WB • p38 MAPK is activated by both inflammatory cytokines and by stress. It is thought to be particularly important in diseases like asthma and autoimmunity but it also plays important roles in the stress response of the nervous system. Like the other MAPKs, p38 is activated by a dual specificity kinase that phosphorylates Thr180 and Tyr182.
Catalog No. 50-288-6724 Supplier Cayman Chemical Supplier No. 100091771 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Thr180 and phospho-Tyr182 of rat p38 MAPK • Host: rabbit • Cross Reactivity: (+) human p38 MAPK • Application(s): WB • p38 MAPK is activated by both inflammatory cytokines and by stress. It is thought to be particularly important in diseases like asthma and autoimmunity but it also plays important roles in the stress response of the nervous system. Like the other MAPKs, p38 is activated by a dual specificity kinase that phosphorylates Thr180 and Tyr182.