Learn More
Eurofins Discoverx p38α/SAPK2a (T106M), active, 250 µg. (14-687M)
Supplier:  Eurofins Discoverx 14687M
Encompass
N-terminal GST-tagged recombinant, human full length p38α/SAPK2a containing the mutation T106M. Activated using DD-MKK6 (14-537) and repurified using glutathione-agarose. Contains the gatekeeper mutation T160M in the ATP-binding domain. This mutation increases resistance to SB203580 as compared to the wild type sequence. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9190