Learn More
Sino Biological Peptide Substrates: p38 Substrate
Catalog No. 502568407
Encompass_Preferred
The synthetic peptide (IPTTPITTTYFFFKKK) contains serine/threonine protein kinase phosphorylation sites and is routinely evaluated as a substrate for p38 family kinases.
Catalog No. 50-256-8407 Supplier Sino Biological Supplier No. P0358
The synthetic peptide (IPTTPITTTYFFFKKK) contains serine/threonine protein kinase phosphorylation sites and is routinely evaluated as a substrate for p38 family kinases.