Learn More
Cayman Chemical p38α MAPK-IN-1, Purity: ≥98%, CAS: 443913-15-3, 10 mg
SDP
Catalog No. 502943413
Encompass_Preferred
A p38α MAPK inhibitor (IC50 = 5,500 nM in an HTRF assay); inhibits LPS-induced TNF-α production in THP-1 cells (EC50 = 6,200 nM); has been used in the study of p38α MAPK inhibitor binding interactions and kinetics
Catalog No. 50-294-3413 Supplier Cayman Chemical Supplier No. 4083910 MG
A p38α MAPK inhibitor (IC50 = 5,500 nM in an HTRF assay); inhibits LPS-induced TNF-α production in THP-1 cells (EC50 = 6,200 nM); has been used in the study of p38α MAPK inhibitor binding interactions and kinetics