Learn More
Carnabio Usa Inc P38B(MAPK11)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 041535
Encompass
p38(MAPK11), Full-length, Wild type, Amino Acid 1-364, NP_002742.3, Expressed in E. coli; 5microg
Catalog No. 50-754-0548