Learn More
Carnabio Usa Inc P38G(MAPK12)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0415510
Encompass
p38(MAPK12), Full-length, Wild type, Amino Acid 1-367, NP_002960.2, Expressed in E. coli; 10microg
Catalog No. 50-754-0910