Learn More
CPC Scientific 5TAMRA-Ile-Pro-Thr-Thr-Pro-Ile-Thr-Thr-Thr-Tyr-Phe-Phe-Phe-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110224
Encompass
Produced in Escherichia coli, this peptide is readily phosphorylated by p38SAPK in in-vitro studies. ONE-LETTER SEQUENCE: 5TAMRA-IPTTPITTTYFFFK-NH2MOLECULAR FORMULA: C109H142N18O24MOLECULAR WEIGHT:2088.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Boros, DL. et al. Clin. Microbial Rev. 2, 250 (1989)
Catalog No. 50-211-0224 Supplier CPC Scientific Supplier No. PROK003B
Produced in Escherichia coli, this peptide is readily phosphorylated by p38SAPK in in-vitro studies. ONE-LETTER SEQUENCE: 5TAMRA-IPTTPITTTYFFFK-NH2MOLECULAR FORMULA: C109H142N18O24MOLECULAR WEIGHT:2088.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Boros, DL. et al. Clin. Microbial Rev. 2, 250 (1989)