Learn More
ATCC P3X63Ag8.653; Plasmacytoma; Mouse (Mus musculus)
Catalog No. 502384613
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
P3X63Ag8.653 is a B lymphoblast cell line that was isolated from a mouse with plasmacytoma. This cell line was deposited by JF Kearney and can be used in immunology and immune system disorder research.
Catalog No. 50-238-4613 Supplier ATCC Supplier No. CRL1580
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
P3X63Ag8.653 is a B lymphoblast cell line that was isolated from a mouse with plasmacytoma. This cell line was deposited by JF Kearney and can be used in immunology and immune system disorder research.