Learn More
ATHENA ES P4-8F™, 500 ml
Catalog No. 5000158182
Encompass_Preferred
BRFF-P4-8FTM is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors designed for culturing immortalized normal prostatic cell lines such as 267-B1.1 P4-8FTM supports the growth of certain established human prostatic cancer cell lines such as PC-3. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.
Catalog No. 50-001-58182 Supplier ATHENA ES Supplier No. 0404
BRFF-P4-8FTM is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors designed for culturing immortalized normal prostatic cell lines such as 267-B1.1 P4-8FTM supports the growth of certain established human prostatic cancer cell lines such as PC-3. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.