Learn More
Enzo Life Sciences Progesterone monoclonal antibody (111.2C7.3) (100µl)
SDP
Supplier:  Enzo Life Sciences ADI905010100
Encompass_Preferred
Clone: 111.2C7.3. Host: Mouse. Isotype: IgG. Immunogen: Progesterone conjugated to bovine serum albumin. Source: Purified from mouse ascites. Species reactivity: Species independent. Applications: ELISA. Purity: Ascites. Formulation: Liquid. Contains sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-6899