Learn More
Lonza Walkersville P4 Primary Cell 96-well Nucleofector Kit
Supplier:  Lonza Walkersville V4SP4096
Encompass
The P4 Primary Cell 96-well Nucleofector Kit is one of our five kits suited for medium-throughput transfection of primary cells when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies for primary cellsCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D-Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10⁴ to 1 x 10⁶ cellsHigh-throughput applications, e.g. s creens of arrayed cDNA, RNAi or CRISPR librariesContent:2.25 mL P4 Primary Cell Nucleofector Solution0.5 mL Supplement 1,50 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)1 96-well Nucleocuvette Plate (20 μl)
Catalog No. NC1026521