Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 8626RBP1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: Polyclonal. Clonality: Polyclonal. Host: Rabbit. Isotype: IgG. Positive Control: HEK293 cells or Prostate Carcinoma or Lung Squamous Cell Carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide (ENNAQTQFSEPQY) corresponding to aa5-17 of human p40. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 3q28. Gene Symbol: TP63. Gene ID: 8626. Ensemble: ENSG00000073282. Uniprot: NP_001316893.
Catalog No. 50-188-5311